missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Barium isopropoxide, 99.5% (metals basis)

Brand:  Alfa Aesar™ 041271.06

Code : NEW

Additional Details : CAS Number : 24363-37-9 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 11339657

  • £104.00 / 5g


Estimated Shipment date
from Supplier 23-05-2018
Add to basket

Specifications

Chemical Identifiers

Barium isopropoxide
Ba[OCH(CH3)2]2
MFCD00050486
24363-37-9
255.52
UN3205

Specifications

Barium isopropoxide
Powder/Lump
(metals basis)
255.52
Moisture Sensitive
White
5g
24363-37-9
99.5%
Ba[OCH(CH3)2]2
MFCD00050486
UN3205
≈200°C (dec.)
Videos
SDS