missing translation for 'onlineSavingsMsg'
Learn More

Barium fluoride, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7050562
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
7787-32-8
BaF2
175.32
MFCD00003450
OYLGJCQECKOTOL-UHFFFAOYSA-L
barium fluoride, baryum fluorure french, baryum fluorure, difluorobarium, barium fluoride, ultra dry, barium ii fluoride, barium fluoride, powder, nickelous sulfate; nickel ii sulfate, barium fluoride trace metals basis
62670
barium(2+) difluoride
[F-].[F-].[Ba++]
This item is not returnable. View return policy
7787-32-8
BaF2
175.32
MFCD00003450
OYLGJCQECKOTOL-UHFFFAOYSA-L
barium fluoride, baryum fluorure french, baryum fluorure, difluorobarium, barium fluoride, ultra dry, barium ii fluoride, barium fluoride, powder, nickelous sulfate; nickel ii sulfate, barium fluoride trace metals basis
62670
barium(2+) difluoride
[F-].[F-].[Ba++]

Barium Fluoride is used in ceramic flux, carbon brushes for electrical equipment, glass making, in the manufacture of other fluorides, crystals for spectroscopy, electronics and dry-film lubricants. Barium Fluoride is used in spectroscopic components. Barium Fluoride is often suitable for applications in the passive IR band (8 to 14 ?m) and is often used as a viewport window for thermography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY
Boiling Point 2260°C
Melting Point 1353°C
Color White
Physical Form Powder
Quantity 10 g
UN Number UN1564
Merck Index 14,974
Solubility Information It is soluble in methanol,ethanol. Sparingly soluble in water.
Formula Weight 175.36
Percent Purity 99.998%
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Density 4.83 g/mL
Assay (metals basis)
Chemical Name or Material Barium fluoride, Ultra dry
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'