missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Barium fluoride crystal optic rectangle, 38.5mm x 19.5mm x 4mm, polished both sides

Brand:  Alfa Aesar™ 038471.KT

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15443655

  • £134.09 / Each


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Specifications

Barium fluoride crystal optic rectangle
38.5mm x 19.5mm x 4mm, Polished both sides
1 each
Videos
SDS