missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium Chloride Dihydrate, Certified AR for Analysis, Fisher Chemical™
57.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
52.95 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.
Use promo code "22914" to get your promotional price.
52.95 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.
Click to view available options
Quantity:
500 g
1 kg
3 kg
5 kg
Description
Barium Chloride Dihydrate, Certified AR for Analysis
Specifications
Boiling Point | 1560°C |
Melting Point | 962°C |
Color | White |
pH | 5 |
Physical Form | Solid |
Quantity | 500 g |
Formula Weight | 244.28g/mol |
Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- Barium chloride dihydrate
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- H301-Toxic if swallowed.
- H332-Harmful if inhaled.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'