missing translation for 'onlineSavingsMsg'
Learn More

Barium chloride, 10% w/v aq. soln., Thermo Scientific Chemicals

Catalog Number p-7050097
24.86 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
500 mL
1 L
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
10361-37-2
BaCl2
208.23
MFCD00003445
WDIHJSXYQDMJHN-UHFFFAOYSA-L
barium chloride, barium dichloride, barium chloride solution, ccris 2286, barium chloride, anhydrous, barium chloride, ultra dry, bariumchlorid, dichlorobarium, dsstox_cid_24508
25204
CHEBI:63317
barium(2+);dichloride
[Cl-].[Cl-].[Ba++]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
10361-37-2
BaCl2
208.23
MFCD00003445
WDIHJSXYQDMJHN-UHFFFAOYSA-L
barium chloride, barium dichloride, barium chloride solution, ccris 2286, barium chloride, anhydrous, barium chloride, ultra dry, bariumchlorid, dichlorobarium, dsstox_cid_24508
25204
CHEBI:63317
barium(2+);dichloride
[Cl-].[Cl-].[Ba++]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Concentration 10% w/v aq. soln.
Formula Weight 208.25
Odor Odorless
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Barium chloride
compliance-icons
Product Identifier
  • Barium chloride, 10% w/v aqueous solution
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Barium chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'