missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Barium chlorate monohydrate, 98%

Brand:  Alfa Aesar™ A10128.36

Code : NEW

Additional Details : CAS Number : 10294-38-9 Weight : 0.50000kg Transport : UN number : 1445 Chem class : 1 Pack group : II

Quantity 500g

 View more versions of this product

Product Code. 11429382

  • £26.80 / 500g


Estimated Shipment date
from Supplier 23-01-2018
Add to basket

Specifications

Chemical Identifiers

Barium chlorate monohydrate
Ba(ClO3)2·H2O
MFCD00149147
10294-38-9
322.26 (304.24 Anhydrous)
UN1445

Specifications

Barium chlorate monohydrate
98%
322.26 (304.24 Anhydrous)
UN1445
10294-38-9
Ba(ClO3)2·H2O
MFCD00149147
500g
Videos
SDS