missing translation for 'onlineSavingsMsg'
Learn More

Barium chloranilate trihydrate, Thermo Scientific Chemicals

Catalog Number p-62219
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
32458-20-1
C6H8BaCl2O7
400.35
MFCD00078327
JFBLKYVZQCFPJE-UHFFFAOYSA-N
barium chloranilate, reag
131877766
barium;2,5-dichloro-3,6-dihydroxycyclohexa-2,5-diene-1,4-dione;trihydrate
O.O.O.[Ba].OC1=C(Cl)C(=O)C(O)=C(Cl)C1=O
This item is not returnable. View return policy
32458-20-1
C6H8BaCl2O7
400.35
MFCD00078327
JFBLKYVZQCFPJE-UHFFFAOYSA-N
barium chloranilate, reag
131877766
barium;2,5-dichloro-3,6-dihydroxycyclohexa-2,5-diene-1,4-dione;trihydrate
O.O.O.[Ba].OC1=C(Cl)C(=O)C(O)=C(Cl)C1=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Brown
Infrared Spectrum Authentic
Packaging Plastic bottle
Quantity 25 g
Solubility Information Solubility in water: soluble
Formula Weight 398.34
Physical Form Powder
Chemical Name or Material Barium chloranilate trihydrate
compliance-icons
Product Identifier
  • Barium chloranilate trihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'