Learn More
Barium calcium copper oxide (2-2-3), 99.9% (metals basis), Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar 039193.09
Code : NEW
Additional Details : Weight : 0.01000kg Transport : UN number : 1564 Chem class : 6.1 Pack group : III
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Barium calcium copper oxide (2-2-3) | |
10 g | |
MFCD00211652 | |
99.9% |
Dry Powder | |
(metals basis) | |
UN1564 |