missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium bromide, anhydrous, 99%, Thermo Scientific Chemicals
43.06 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
50 g
250 g
Description
Barium bromide, anhydrous is used as the chemical intermediate, and the spice cosmetics chemical additive. It is also used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 850°C |
Color | White |
Physical Form | Powder |
Quantity | 50 g |
UN Number | UN1564 |
Merck Index | 14,968 |
Solubility Information | Soluble in water |
Formula Weight | 297.14 |
Percent Purity | 99% |
Odor | Odorless |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'