missing translation for 'onlineSavingsMsg'
Learn More

Barium aluminum oxide, tech., Thermo Scientific Chemicals

Catalog Number p-7049778
Click to view available options
Quantity:
10 g
100 g
500 g
This item is not returnable. View return policy
12004-04-5
MFCD00049716
Barium aluminate
This item is not returnable. View return policy
12004-04-5
MFCD00049716
Barium aluminate

Barium aluminum oxide is used for getter alloys in vacuum tubes. It also finds application as deoxidizer for copper, Frary’s metal, lubricant for anode rotors in x-ray tubes and spark-plug alloys. It is used to prepare heat-resistant refractory and super-refractory concretes. Further, it gives a better protection against gamma radiation and X-rays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Powder
Quantity 10 g
UN Number UN1564
Solubility Information Insoluble in water.
Formula Weight 561.98
Grade Technical
Odor Odorless
Chemical Name or Material Barium aluminum oxide
compliance-icons
Product Identifier
  • Barium aluminum oxide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P330-Rinse mouth.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'