Learn More
Thermo Scientific Alfa Aesar Barium 2-methoxyethoxide, 99.5% (metals basis), 10% w/v in 2-methoxyethanol
Brand: Thermo Scientific Alfa Aesar 041256.AE
Barium 2-methoxyethoxide, 99.5% (metals basis), 10% w/v in 2-methoxyethanol
Code : NEW
Additional Details : CAS Number : 115503-13-4 Weight : 0.01000kg Transport : UN number : 1188 Chem class : 3 Pack group : III