missing translation for 'onlineSavingsMsg'
Learn More

Bare Nichrome Wire

125g per reel.

Brand:  Edulab PH1064A

UNSPSC : 12000000

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15287818

  • £203.00 / Pack of 10


Estimated Shipment
04-08-2017
Add to basket

Specifications Description & Specifications

Specifications

15.35 x 5.9 x 3.9 in. (39 x 15 x 10cm)
18 ga
15cm
39cm
1.56kg
10cm
0.13cm
Nichrome Wire