missing translation for 'onlineSavingsMsg'
Learn More

Barbituric acid, 99+%, Thermo Scientific Chemicals

Catalog Number p-7203421
23.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
Regulatory: The product is controlled by Misuse of Drugs Act 1971 and the Misuse of Drugs Regulations 2001, all customers must complete a declaration before delivery.
67-52-7
C4H4N2O3
128.09
MFCD00006666
HNYOPLTXPVRDBG-UHFFFAOYSA-N
barbituric acid, malonylurea, 2,4,6 1h,3h,5h-pyrimidinetrione, 6-hydroxyuracil, pyrimidinetrione, 2,4,6-trihydroxypyrimidine, 2,4,6-pyrimidinetrione, pyrimidine-2,4,6 1h,3h,5h-trione, barbiturate, malonylharnstoff
6211
CHEBI:16294
1,3-diazinane-2,4,6-trione
O=C1CC(=O)NC(=O)N1
This item is not returnable. View return policy
Regulatory: The product is controlled by Misuse of Drugs Act 1971 and the Misuse of Drugs Regulations 2001, all customers must complete a declaration before delivery.
67-52-7
C4H4N2O3
128.09
MFCD00006666
HNYOPLTXPVRDBG-UHFFFAOYSA-N
barbituric acid, malonylurea, 2,4,6 1h,3h,5h-pyrimidinetrione, 6-hydroxyuracil, pyrimidinetrione, 2,4,6-trihydroxypyrimidine, 2,4,6-pyrimidinetrione, pyrimidine-2,4,6 1h,3h,5h-trione, barbiturate, malonylharnstoff
6211
CHEBI:16294
1,3-diazinane-2,4,6-trione
O=C1CC(=O)NC(=O)N1

Barbituric acid, CAS # 67-52-7, (synonym: malonylurea) is the parent compound used in the production of barbiturates that act as sedative drugs. It is also used extensively in the manufacturing of plastics, textiles, and polymers.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

• Barbituric acid is a white, odorless, water-soluble crystalline powder
• It is a strong acid in an aqueous medium


Applications

• Barbituric acid uses the Knoevenegal condensation reaction to be converted into barbiturate drugs that acts as central nervous system depressants
• It is an active ingredient in the synthesis of Vitamin B12
• It is also used in electrochemical oxidation of iodine using cyclic voltammetry and controlled potential coulometry

TRUSTED_SUSTAINABILITY
Assay Percent Range 99+%
Merck Index 15, 958
Formula Weight 128.09
Quantity 100 g
Packaging Plastic bottle
Chemical Name or Material Barbituric acid, 99+%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'