Learn More
Masterflex™ Barbed Fitting Reducing Connector
Ideal for low-pressure applications
£19.12 - £19.53
Specifications
Description Barb
Material Polypropylene
Sterilization Method Ethylene Oxide, Gamma
View More Specs
Products 2
Product Code Brand Tubing Size Price Quantity & Availability  
Product Code Brand Tubing Size Price Quantity & Availability  
13470899
View Documents
Masterflex™
40610-20
1 in. x 1/2 in. ID, 11/32 in., 3-5/8 in. £19.12
Pack of 5
Qty Price Break Available View alternative products
Save Up to 
»
Estimated Shipment
18-12-2020
Add to basket
13440909
View Documents
Masterflex™
40610-10
1/2 in. x 1/4 in. ID, 1/8 in., 2-15/32 in. £19.53
Pack of 10
Qty Price Break Available View alternative products
Save Up to 
»
Estimated Shipment
15-12-2020
Add to basket
Description
Description
  • These economical barbed fittings easily connect fluid paths without the use of tools.
  • Just slip your flexible tubing or hose onto the barb to create a secure seal
    Ideal for low-pressure applications.
  • Chemically resistant.
  • Available in a variety of sizes, materials, and configurations.
Specifications
Specifications
Barb
Polypropylene
Ethylene Oxide, Gamma
SDS