Learn More
Masterflex™ Barbed Fitting Reducing Connector
Ideal for low-pressure applications
Brand: Masterflex™ 40610-10
Code : NEW
Additional Details : Weight : 0.05000kg
Description
Ideal for low-pressure applications
- These economical barbed fittings easily connect fluid paths without the use of tools.
- Just slip your flexible tubing or hose onto the barb to create a secure seal
Ideal for low-pressure applications. - Chemically resistant.
- Available in a variety of sizes, materials, and configurations.
Specifications
Barbed Fittings, Reducing Connector, Natural PP, 1/2 in. x 1/4 in. ID, 1/8 in., 2-15/32 in. | |
Polypropylene | |
Ethylene Oxide, Gamma |
Barb | |
1/2 in. x 1/4 in. ID, 1/8 in., 2-15/32 in. | |
Natural |