Learn More
Tocris Bioscience™ BAN ORL 24
Potent and selective NOP antagonist
Brand:  Tocris Bioscience™ 366110
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13443374
  • £210.21 / 10mg
Estimated Shipment date
from Supplier 17-12-2019
Description
Description
Potent and selective NOP receptor antagonist (IC50 values are 0.27, 2500, 6700 and > 10000 nM for NOP, κ-, μ- and δ-receptors respectively). Inhibits nociceptin-induced stimulation of [35S]-GTPγS binding and Ca2+ mobilization in CHO cells in vitro and antagonizes NOP agonist-induced reduction in locomotor activity in vivo. Brain penetrant.
Specifications
Specifications
BAN ORL 24
>97
10mg
BAN ORL 24
506.51