Learn More
Tocris Bioscience™ BAN ORL 24
Potent and selective NOP antagonist
Brand:  Tocris Bioscience™ 366150
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13463374
  • £710.36 / 50mg
Estimated Shipment
29-03-2019
Add to basket
Description
Description
Potent and selective NOP receptor antagonist (IC 50 values are 0.27, 2500, 6700 and > 10000 nM for NOP, κ-, μ- and δ-receptors respectively). Inhibits nociceptin-induced stimulation of [ 35 S]-GTPγS binding and Ca 2+ mobilization in CHO cells in vitro and antagonizes NOP agonist-induced reduction in locomotor activity in vivo. Brain penetrant.
Specifications
Specifications
BAN ORL 24
>97
50mg
BAN ORL 24
506.51