missing translation for 'onlineSavingsMsg'
Learn More

J.T. Baker Baker-flex™, Cellulose PEI-F, J.T. Baker™

Catalog Number 15527594
973.75 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.


Click to view available options
Quantity:
25 Sheets
This item is not returnable. View return policy
9004-34-6
This item is not returnable. View return policy
9004-34-6

Flexible TLC Sheet

Color White
Physical Form Crystal or Powder
Solubility Information Insoluble in water
Quantity 25 Sheets
Odor Odorless
Chemical Name or Material Cellulose PEI-F
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'