missing translation for 'onlineSavingsMsg'
Learn More

J.T. Baker Baker-flex™, Aluminum Oxide IB-F, J.T. Baker™

Catalog Number p-5548222
643.93 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.


Click to view available options
Quantity:
200 Sheets
This item is not returnable. View return policy
1344-28-1
Al2O3
101.96
MFCD00003424
PNEYBMLMFCGWSK-UHFFFAOYSA-N
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
9989226
[O--].[O--].[O--].[Al+3].[Al+3]
This item is not returnable. View return policy
1344-28-1
Al2O3
101.96
MFCD00003424
PNEYBMLMFCGWSK-UHFFFAOYSA-N
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
9989226
[O--].[O--].[O--].[Al+3].[Al+3]

Flexible TLC Sheet

Quantity 200 Sheets
Solubility Information Insoluble in water
IUPAC Name dialuminum;oxygen(2-)
Odor Odorless
Specific Gravity 1L = 4.00kg
Chemical Name or Material Aluminum Oxide IB-F
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'