missing translation for 'onlineSavingsMsg'
Learn More

Baculovirus Envelope gp64, clone: AcV5, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-6995-81

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15267967

  • £81.42 / 50µg


Call for latest availability
+44 (0)1509 555 500
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The AcV5 antibody reacts with the gp64 envelope protein of the baculovirus Autographa californica (AcMNPV).Applications Reported: The AcV5 antibody has been reported for use in immunoblotting (WB). AcV5 can be used in identifying virally-infected insect cells and biochemical analysis of the gp64 protein.Applications Tested: The AcV5 antibody has been tested by immunoblotting (WB) of baculovirus infected insect cells. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

baculovirus envelope gp64 protein
AcV5
Unconjugated
Purified
Murine
50μg
Store at 2-8°C.
Monoclonal
Western Blot
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
BV gp64
IgG2b
RUO
Primary
Videos