missing translation for 'onlineSavingsMsg'
Learn More

Baculovirus Envelope gp64, clone: AcV1, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-6991-81

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15207967

  • £56.31 / 50µg


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Description: The AcV1 monoclonal antibody reacts with the gp64 envelope protein of the baculovirus Autographa californica (AcMNPV). The gp64 envelope protein is essential for virus infectivity and is expressed on the surface of baculovirus-infected cells within six hours of infection. The AcV1 antibody can be used in flow cytometry-based viral titration experiments.Applications Reported: The AcV1 antibody has been reported for use in flow cytometric analysis. This antibody can be used in identifying virally-infected insect cells.Applications Tested: The AcV1 antibody has been tested by flow cytometry on baculovirus infected insect cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

baculovirus envelope gp64 protein
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
BV gp64
IgG2a
50μg
Store at 2-8°C.
Primary
Flow Cytometry
AcV1
Unconjugated
Purified
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Videos

For Research Use Only.