missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bacteriological Peptone, Ultrapure, Thermo Scientific™
High grade peptone prepared specifically for general bacteriological purposes.
£38.58 - £644.29
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15895388
|
Thermo Scientific
J20048.TD |
1 lb. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15805398
|
Thermo Scientific
J20048.=A |
5 lb. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15815398
|
Thermo Scientific
J20048.5 |
25 lb. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Format: Powder
- CAS: 73049-73-7
- MDL Number: MFCD00131829
RUO – Research Use Only