Learn More
Technical Service Consultants™ Bacterial Preservation System PP Freezer Rack Box
Facilitate the integrity of your bacterial preservation system using the Technical Service Consultants™ Bacterial Preservation System PP Freezer Rack Box. This system enables you to reduce the labor costs of continual sub-culturing and eliminates the need to purchase expensive freeze drying equipment. The culture viability can be checked regularly without sacrificing the entire vial.
Brand:  Technical Service Consultants™ TS71-YE
Code : 03
Additional Details : Weight : 0.76140kg
 View more versions of this product
Product Code. 12867745
  • £203.00 / Pack of 100
Estimated Shipment
19-06-2018
Add to basket
Description
Description
  • 100-vial capacity - available in range of colors
  • Contains 20 to 25 treated beads in cryopreservation fluid
  • Long-term storage of::bacteria, fungi and yeasts; quality control cultures; culture collections; and cross-infection isolates
Specifications
Specifications
Yellow
-20 to -70°C
Holds 100 Vials
Porous ceramic beads suspended in a cryopreservative and contained in a freezer vial
Videos
Documentation
Technical Documents
View accessory products
Special Offers
Special Offers
  • Use code 13239 to receive this promotion.
    : 13239
    2018-08-31
    *Discount will apply to qualifying orders received by Fisher Scientific between 15/05/2018 and 31/08/2018 subject to availability of the product in stock. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy or if the product is no longer available in stock. Other restrictions may apply.
    Terms & Conditions