missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Bactenecin, Bovine

For cellular and molecular biology applications

Brand:  Alfa Aesar™ J66855.MB

Code : NEW

Additional Details : Weight : 0.00025kg

 View more versions of this product

Product Code. 15418389

  • £542.90 / 25mg


Estimated Shipment date
from Supplier 10-05-2018
Add to basket

Specifications

Specifications

1483.89
Bovine
Lyophilized powder
RUO
Bactenecin, bovine
HPLC
25mg
Arg-Leu-Cys-Arg-Ile-Val-Val-Ile-Arg-Val-Cys-Arg (Disulfide bridge: Cys3-Cys11)
-30° to -10°C
Videos
SDS

For Research Use Only