missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bacitracin, potency min. 60 Units/mg

Catalog Number p-62190
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
1405-87-4
C66H103N17O16S
MFCD00062640
This item is not returnable. View return policy
1405-87-4
C66H103N17O16S
MFCD00062640

Bacitracin, CAS # 1405-87-4, is cyclic polypeptide-based antibiotic which exhibits activity against microbes causing pneumonia, skin, and eye infections.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Bacitracin is a mixture of at least nine different cyclic polypeptides, originally isolated from Bacillus bacteria in 1945

• Bacitracins work by binding divalent metal cations and preventing the hydrolysis of lipids into usable cell wall components

Applications

• Bacitracin has been paired with nanoparticles in some laboratory studies to increase its effectivity

• It is suitable for the laboratory study of bacterial resistance, infection, and cross-reactivity

TRUSTED_SUSTAINABILITY
Type Bacitracin
Physical Form Powder
Melting Point 221.0°C to 225.0°C
Infrared Spectrum Authentic
Loss on Drying 5.0% max. (1 g, 60°C, 3 Hrs) (P2O5) (not exceeding 0.1 kPa)
Merck Index 15, 927
Packaging Glass Bottle
Solubility Information Solubility in water: soluble. Other solubilities: soluble in glacial acetic acid, methanol and, alcohol, practically insoluble in ether, chloroform and, acetone
Formula Weight 1422.71
Percent Purity 60 I.U./mg min. (potency, on dried subst.)
Color Beige or White
Concentration ≥60 I.U./mg (potency, on dried subst.)
Quantity 5 g
Chemical Name or Material Bacitracin
Show More Show Less
compliance-icons
Product Identifier
  • Bacitracin
Signal Word
  • Warning
Hazard Category
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'