missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BACE-2 Ectodomain Sheep anti-Human, Polyclonal, R&D Systems™
Sheep Polyclonal Antibody has been used in 1 publication
POA
Specifications
Antigen | BACE-2 |
---|---|
Immunogen | Mouse myeloma cell line NS0-derived recombinant human BACE-2 Ectodomain Gly63-Pro466 Accession # Q9Y5Z0 |
Host Species | Sheep |
Primary or Secondary | Primary |
Classification | Polyclonal |
Description
BACE-2 Polyclonal specifically detects BACE-2 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
BACE-2 | |
Sheep | |
Polyclonal | |
IgG | |
AEPLC, Asp 1, ASP1, ASP21, Aspartic-like protease 56 kDa, Aspartyl protease 1, BACE2, beta secretase 2, beta-secretase 2, beta-site amyloid beta A4 precursor protein-cleaving enzyme 2, Beta-site amyloid precursor protein cleaving enzyme 2, Beta-site APP cleaving enzyme 2, beta-site APP-cleaving enzyme 2, beta-site APP-cleaving enzyme 2, EC 3.4.2310ALP5656 kDa aspartic-like protease, CEAP1, Down region aspartic protease, Down syndrome region aspartic protease, DRAPBAE2, EC 3.4.23, EC 3.4.23.45, Memapsin-1, Membrane-associated aspartic protease 1, Theta-secretase, transmembrane aspartic proteinase Asp1 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Western Blot, Immunoprecipitation | |
RUO |
Mouse myeloma cell line NS0-derived recombinant human BACE-2 Ectodomain Gly63-Pro466 Accession # Q9Y5Z0 | |
Primary | |
Human | |
Unconjugated | |
25825 | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only