missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat B7-1/CD80 Antibody, R&D Systems™
Mouse Monoclonal Antibody
POA
Specifications
Antigen | B7-1/CD80 |
---|---|
Clone | 210408 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
B7-1/CD80 Monoclonal specifically detects B7-1/CD80 in Rat samples. It is validated for Western Blot.Specifications
B7-1/CD80 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Rat | |
AAB60503 | |
941 | |
Mouse myeloma cell line NS0-derived recombinant rat B7-1/CD80 Ile39-Gln248 Accession # AAB60503 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects rat B7-1/CD80 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human B7-1, recombinant mouse (rm) B7-1, rmB7-2, recombinant rat B7-2, rmB7-H1, rmB7-H2, rmB7-H3, or rmPD-L2 is observed. |
210408 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Activation B7-1 antigen, B7, B71, BB1, CD28LG1, CD28LGB7-1 antigen), CD80, CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80 | |
CD80 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only