missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B7-1/CD80 Goat anti-Rat, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
POA
Specifications
Antigen | B7-1/CD80 |
---|---|
Immunogen | Mouse myeloma cell line NS0-derived recombinant rat B7-1/CD80 Ile39-Gln248 Accession # AAB60503 |
Host Species | Goat |
Primary or Secondary | Primary |
Classification | Polyclonal |
Description
B7-1/CD80 Polyclonal specifically detects B7-1/CD80 in Rat samples. It is validated for Western Blot, Immunocytochemistry.Specifications
B7-1/CD80 | |
Goat | |
Polyclonal | |
IgG | |
Activation B7-1 antigen, B7, B71, BB1, CD28LG1, CD28LGB7-1 antigen), CD80, CD80 antigen, CD80 molecule, costimulatory factor CD80, costimulatory molecule variant IgV-CD80, CTLA-4 counter-receptor B7.1, T-lymphocyte activation antigen CD80 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Western Blot, Immunocytochemistry | |
RUO |
Mouse myeloma cell line NS0-derived recombinant rat B7-1/CD80 Ile39-Gln248 Accession # AAB60503 | |
Primary | |
Rat | |
Unconjugated | |
941 | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only