Learn More
B3GNT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA553307
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: EEQDQYFEFF PPSPRSVDQV KAQLRTALAS GGVLDASGDY RVYRGLLKTT MDPNDVILAT HASVDNLLHL SGLLERWEGP LSVSVFAATK EEAQLATVLA YALSSHCPDM RARVAMHLVC PSRYEAAVPD PREPGEFALL RSCQ Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 97%.
B3GNT1 Polyclonal antibody specifically detects B3GNT1 in Human samples. It is validated for Immunohistochemistry.Specifications
B3GNT1 | |
Polyclonal | |
Unconjugated | |
B4GAT1 | |
1500032M01Rik; B3GNT1; B3GN-T1; B3GNT1 (HGNC:15685); B3gnt6; B4GAT1; beta-1,3-N-acetylglucosaminyltransferase bGnT-6; beta-1,4-glucuronyltransferase 1; BETA3GNT1; BETA3GNTI; EC 2.4.1.149; I-beta-1,3-N-acetylglucosaminyltransferase; iGAT; iGNT; MDDGA13; N-acetyllactosaminide beta-1,3-N-acetylglucosaminyltransferase; Poly-N-acetyllactosamine extension enzyme; UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 1; UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11041 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
Lot-Specific | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O43505 | |
B4GAT1 | |
Recombinant protein corresponding to Human B3GNT1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only