Learn More
B3GAT3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA562383
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: AASGLLFTHL VVLTPKAQRL REGEPGWVHP RGVEQRNKAL DWLRGRGGAV GGEKDPPPPG TQGVVY Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 94%.
B3GAT3 Polyclonal antibody specifically detects B3GAT3 in Human samples. It is validated for Immunohistochemistry.Specifications
B3GAT3 | |
Polyclonal | |
Unconjugated | |
B3GAT3 | |
2810405M13Rik; 3-beta-glucuronosyltransferase; B3GAT3; Beta-1,3-glucuronyltransferase 3; beta-1,3-glucuronyltransferase 3 (glucuronosyltransferase I); galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 3; GLCATI; GlcAT-I; glcUAT-I; glucuronosyltransferase I; glucuronosyltransferase-I; glucuronyltransferase I; JDSCD; LOW QUALITY PROTEIN: galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 3; Sqv-8-like protein; UDP-GlcUA:Gal beta-1,3-Gal-R glucuronyltransferase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26229 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O94766 | |
B3GAT3 | |
Recombinant protein corresponding to Human B3GAT3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only