missing translation for 'onlineSavingsMsg'
Learn More

B-HT 933 dihydrochloride, Tocris Bioscience™

Selective α2 agonist

Brand:  Tocris (Bio-Techne) 275810

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13440534

  • £170.28 / 10mg

Estimated Shipment
28-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective α2-adrenoceptor agonist. Exhibits greater than 300-fold selectivity for the α2-adrenoceptor over the α1-adrenoceptor.
Specifications

Chemical Identifiers

36067-72-8
254.155
b-ht 933 dihydrochloride, azepexole hydrochloride, azepexole dihydrochloride, 2-amino-6-ethyl-4,5,7,8-tetrahydro-6h-oxazolo 5,4-d azepine dihydrochloride, 5,6,7,8-tetrahydro-6-ethyl-4h-oxazolo 4,5-d azepin-2-amine dihydrochloride, 6-ethyl-5,6,7,8-tetrahydro-4h-oxazolo 4,5-d azepin-2-amine dihydrochloride, dsstox_cid_25756, dsstox_rid_81099, dsstox_gsid_45756, azepexole hcl
6-ethyl-4,5,7,8-tetrahydro-[1,3]oxazolo[4,5-d]azepin-2-amine;dihydrochloride
C9H17Cl2N3O
HBLPYIOKPJVFQW-UHFFFAOYSA-N
169743
CCN1CCC2=C(CC1)OC(=N2)N.Cl.Cl

Specifications

B-HT 933 dihydrochloride
36067-72-8
b-ht 933 dihydrochloride, azepexole hydrochloride, azepexole dihydrochloride, 2-amino-6-ethyl-4,5,7,8-tetrahydro-6h-oxazolo 5,4-d azepine dihydrochloride, 5,6,7,8-tetrahydro-6-ethyl-4h-oxazolo 4,5-d azepin-2-amine dihydrochloride, 6-ethyl-5,6,7,8-tetrahydro-4h-oxazolo 4,5-d azepin-2-amine dihydrochloride, dsstox_cid_25756, dsstox_rid_81099, dsstox_gsid_45756, azepexole hcl
CCN1CCC2=C(CC1)OC(=N2)N.Cl.Cl
254.155
254.16
10mg
C9H17Cl2N3O
HBLPYIOKPJVFQW-UHFFFAOYSA-N
6-ethyl-4,5,7,8-tetrahydro-[1,3]oxazolo[4,5-d]azepin-2-amine;dihydrochloride
169743
>99%
Videos