missing translation for 'onlineSavingsMsg'
Learn More

B-HT 920, Tocris Bioscience™

D2 receptor agonist. Also α2 agonist and 5-HT3 antagonist

Brand:  Tocris (Bio-Techne) 275910

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13460534

  • £128.25 / 10mg


Estimated Shipment
21-02-2019
Add to basket

Description

Description

Dopamine D2 receptor agonist, α2-adrenoceptor agonist and 5-HT3 receptor antagonist. Displays antiParkinsonian activity.
Specifications

Chemical Identifiers

36085-73-1
282.227
talipexole dihydrochloride, b-ht 920 dihydrochloride, unii-9r6e1d8h1o, 6-allyl-2-amino-5,6,7,8-tetrahydro-4h-thiazolo 4,5-d azepine dihydrochloride, 2-amino-6-allyl-4,5,7,8-tetrahydro-6h-thiazolo 5,4-d azepin-dihydrochlorid german, 4h-thiazolo 4,5-d azepin-2-amine, 5,6,7,8-tetrahydro-6-2-propenyl-, dihydrochloride, 5,6,7,8-tetrahydro-6-2-propenyl-4h-thiazolo 4,5-d azepin-2-amine dihydrochloride, domin tn, pubchem19117, 5,6,7,8-tetrahydro-6-2-propenyl-4h-thiazolo 4,5-d azepine-2-amine dihydrochloride
6-prop-2-enyl-4,5,7,8-tetrahydro-[1,3]thiazolo[4,5-d]azepin-2-amine;dihydrochloride
C10H17Cl2N3S
DPQAXNSOFFYKDS-UHFFFAOYSA-N
104870
C=CCN1CCC2=C(CC1)SC(=N2)N.Cl.Cl

Specifications

B-HT 920
36085-73-1
talipexole dihydrochloride, b-ht 920 dihydrochloride, unii-9r6e1d8h1o, 6-allyl-2-amino-5,6,7,8-tetrahydro-4h-thiazolo 4,5-d azepine dihydrochloride, 2-amino-6-allyl-4,5,7,8-tetrahydro-6h-thiazolo 5,4-d azepin-dihydrochlorid german, 4h-thiazolo 4,5-d azepin-2-amine, 5,6,7,8-tetrahydro-6-2-propenyl-, dihydrochloride, 5,6,7,8-tetrahydro-6-2-propenyl-4h-thiazolo 4,5-d azepin-2-amine dihydrochloride, domin tn, pubchem19117, 5,6,7,8-tetrahydro-6-2-propenyl-4h-thiazolo 4,5-d azepine-2-amine dihydrochloride
C=CCN1CCC2=C(CC1)SC(=N2)N.Cl.Cl
282.227
282.23
10mg
C10H17Cl2N3S
DPQAXNSOFFYKDS-UHFFFAOYSA-N
6-prop-2-enyl-4,5,7,8-tetrahydro-[1,3]thiazolo[4,5-d]azepin-2-amine;dihydrochloride
104870
>98%
Videos