missing translation for 'onlineSavingsMsg'
Learn More

BUCHI™ V-850 and V-855 Vacuum Controllers

Universal units for the display, adjustment and control of all vacuum systems

Brand:  BUCHI™ 047298

Code : 61

Additional Details : Weight : 0.84000kg

 View more versions of this product

Product Code. 10155172

  • £2452.33 / Each


Estimated Shipment
27-06-2018
Add to basket

Specifications

Specifications

12cm
120mm
100/230V
160mm
30VDC (with connection to Rotavapor and Vacuum Pump V-700/710) or 85 to 264VDC power pack
V-855 Vacuum Controller
+10 to +40°C
106mm
10° to 40°C
540g
CE, CSA, UL
Rotavapor R-210/215 and V-700/V-710 vacuum pump
Support, Communication Cable
1400 to 0mbar (hPa);, 1050 to 1 torr
Videos
Documentation View accessory products