missing translation for 'onlineSavingsMsg'
Learn More

BUCHI™ V-710 Vacuum Pump Systems

Oil-free, chemically resistant vacuum pump with high flowrates and low ultimate vacuum ideal for wet chemistry

Brand:  BUCHI™ 072412

Code : 61

Additional Details : Weight : 13.79000kg

 View more versions of this product

Product Code. 10380423

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

100/240V 50/60Hz
Laboratory and industrial Rotavapor™
V-710
10° to 40°C
10.4kg
100/240V
29.7cm
Power cable, sound absorber, vacuum tubing and connections and operating instructions, woulff bottle, secondary cold trap, V-801 EasyVac vacuum controller
1600rpm
3.1m3/Hour
PEEK, PTFE, Glass, FEP, FFKM
41 to 55dBA
370w
Brushless DC
50/60Hz
37.8cm
40°C
CE, CSA
Videos
Documentation