missing translation for 'onlineSavingsMsg'
Learn More

V-710 Vacuum Pump Systems

Oil-free, chemically resistant vacuum pump with high flowrates and low ultimate vacuum ideal for wet chemistry

Brand:  Buchi 072012

Code : 61

Additional Details : Weight : 13.22000kg

 View more versions of this product

Product Code. 12796914

  • £4863.01 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

Brushless DC
Power cable, sound absorber, vacuum tubing and connections and operating instructions, woulff bottle, secondary cold trap
CE, CSA
PEEK, PTFE, Glass, FEP, FFKM
41 to 55dBA
10.4kg
10°C
Laboratory and industrial Rotavapor™
11.68 in.
14.87 in.
6.96 in.
100/240V
1600rpm
100/240V 50/60Hz
V-710
370w
40°C
3.1m³/hr.
50/60Hz
29.7cm
37.8cm
Videos
Documentation