Learn More
BUCHI Flask Seals for Industrial Rotavapor™
Original BUCHI seal for Industrial Rotavapor™. Optimize the performance of your distillation with high quality and chemically resistant replacement parts.
£141.00 - £815.00
Specifications
For Use With (Equipment) Flask seals
View More Specs
Products 2
Product Code Brand Type Price Quantity & Availability  
Product Code Brand Type Price Quantity & Availability  
10528045
View Documents Promotion Details
BUCHI
041121
Evaporating Flask Seal, Complete
£815.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 20-05-2024
to see stock.
Add to basket
10270402
View Documents Promotion Details
BUCHI
034152
Beaker Flask Seal
£141.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 21-05-2024
to see stock.
Add to basket
Description
Description
  • High chemical resistance
  • OEM seal for proper fit and performance
  • PTFE, Viton and silicone seal
Specifications
Specifications
Flask seals