missing translation for 'onlineSavingsMsg'
Learn More

AZM 475271, Tocris Bioscience™

Src tyrosine kinase inhibitor

Brand:  Tocris (Bio-Techne) 396350

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13463894

  • £1021.65 / 50mg

Estimated Shipment date
from Supplier 26-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Src tyrosine kinase inhibitor. Reduces tumor size, vascularity and metastasis, and increases apoptosis in human pancreatic cells grown in nude mice. Also exhibits antiangiogenic activity in vitro and in vivo. Sensitizes tumor cells to the cytotoxic effects of gemcitabine (Cat. No. 3259).
Specifications

Chemical Identifiers

476159-98-5
442.944
n-2-chloro-5-methoxyphenyl-6-methoxy-7-1-methylpiperidin-4-yl methoxy quinazolin-4-amine, methoxy aniline 7, azd0530 analogue 35, 4-quinazolinamine, n-2-chloro-5-methoxyphenyl-6-methoxy-7-1-methyl-4-piperidinyl methoxy, n-2-chloro-5-methoxyphenyl-6-methoxy
N-(2-chloro-5-methoxyphenyl)-6-methoxy-7-[(1-methylpiperidin-4-yl)methoxy]quinazolin-4-amine
C23H27ClN4O3
WPOXAFXHRJYEIC-UHFFFAOYSA-N
5330175
CN1CCC(CC1)COC2=C(C=C3C(=C2)N=CN=C3NC4=C(C=CC(=C4)OC)Cl)OC

Specifications

50mg
476159-98-5
n-2-chloro-5-methoxyphenyl-6-methoxy-7-1-methylpiperidin-4-yl methoxy quinazolin-4-amine, methoxy aniline 7, azd0530 analogue 35, 4-quinazolinamine, n-2-chloro-5-methoxyphenyl-6-methoxy-7-1-methyl-4-piperidinyl methoxy, n-2-chloro-5-methoxyphenyl-6-methoxy
CN1CCC(CC1)COC2=C(C=C3C(=C2)N=CN=C3NC4=C(C=CC(=C4)OC)Cl)OC
442.944
442.94
AZM 475271
C23H27ClN4O3
WPOXAFXHRJYEIC-UHFFFAOYSA-N
N-(2-chloro-5-methoxyphenyl)-6-methoxy-7-[(1-methylpiperidin-4-yl)methoxy]quinazolin-4-amine
5330175
>99%
Videos
Documents