Learn More
Dynalon™ Azlon™ Solvent Venting Wash Bottles
Description
Translucent low density polyethylene wash bottles feature unique two-way valve that stops vapor pressure build-up and eliminates the hazard of volatile solvent leaks. Wide neck provides for easier and safer filling. Plus, for quick identification, cadmium-free polypropylene dispensing closures are color coded. And, for optimum control in dispensing, the jet is precision molded. Each bottle features a clear, indelible label describing the contents, international hazard codes and instructional diagrams for safe usage. Unlabeled bottles with write-on label areas and complete set of adhesive hazard labels for custom use are also available. Supplied in convenient shelf packs of five.
Order Info
Bottles are available in 250 or 500mL capacities with five common solvent labels.
Compliance
All bottles have SPI recycling code stamped on base.
Specifications
Capacity | 8 oz., 250 mL |
Closure Material | Polypropylene |
Height (English) | 8 in. |
Height (Metric) | 203.2 mm |
Width (English) | 2.25 in. |
Capacity (Metric) | 250 mL |
Capacity (English) | 8 oz. |
Color | Translucent |
For Use With (Application) | Methanol |
Material | LDPE |
Show More |