missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azlon™ Polypropylene Autoclave bags
Autoclave waste in this blue autoclave bag for biohazardous material. The Azlon™ polypropylene autoclave bag has printed instructions and can be autoclaved at temperatures up to 121 degrees C.
Brand: Azlon BWY204
Code : 02
Additional Details : Weight : 3.66000kg
Description
- Suitable for autoclaving biohazardous waste
- Strong side weld construction (50μm)
- Printed with instructions for use and biohazard symbol
Specifications
Autoclavable | |
Polypropylene | |
896 mm |
Blue | |
608 mm |