missing translation for 'onlineSavingsMsg'
Learn More

Azlon™ Pivot Ring Magnetic Stirrer Bar

Constructed of magnetic PTFE. Azlon™ Pivot Ring Stirrer Bar includes a central pivot ring that ensures efficient spinning even on curved or uneven bases.

Brand:  Azlon SWN503

Code : A0

Additional Details : Weight : 0.09000kg

 View more versions of this product

Product Code. 10585105

  • £5.55 / Pack of 2


In Stock 26
Add to basket

Description

Description

  • Constructed of magnetic PTFE
  • Central pivot ring ensures efficient spinning even on curved or uneven bases
Specifications

Specifications

Yes
0.78 Lin.
Stirrer bar
20mm
Pivot ring
PTFE
2/Pk.
6mm
Magnet
Videos
Documentation

Technical Documents