missing translation for 'onlineSavingsMsg'
Learn More

Azetidine, 98%, Thermo Scientific Chemicals

Catalog Number 10481681
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
503-29-7
C3H7N
57.09
HONIICLYMWZJFZ-UHFFFAOYSA-N
azacyclobutane, trimethylene imine, 1,3-propylenimine, trimethylenimine, azetidin, unii-37s883xdwr, azetidine, l, polypropylene amine, azete, tetrahydro, azetidine
10422
CHEBI:30968
azetidine
C1CNC1
This item is not returnable. View return policy
503-29-7
C3H7N
57.09
HONIICLYMWZJFZ-UHFFFAOYSA-N
azacyclobutane, trimethylene imine, 1,3-propylenimine, trimethylenimine, azetidin, unii-37s883xdwr, azetidine, l, polypropylene amine, azete, tetrahydro, azetidine
10422
CHEBI:30968
azetidine
C1CNC1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Density 0.8500g/mL
Boiling Point 61.0°C to 62.0°C
Flash Point −20°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4290 to 1.4330
Quantity 1 g
Beilstein 20,2
Specific Gravity 0.85
Solubility Information Solubility in water: miscible.
Formula Weight 57.09
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Azetidine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Azetidine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'