missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Azadibenzocyclooctyne-PEG4-NHS ester

Brand:  Alfa Aesar™ J64126.MB

Code : NEW

Additional Details : Weight : 0.00025kg

Quantity 25mg

 View more versions of this product

Product Code. 15406219

  • £174.90 / 25mg


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Description

Description

Applications
Azadibenzocyclooctyne-PEG4-NHS ester is applied for protein-peptide conjugation, peptide-small molecule conjugation, 18F radiolabeling, protein-oligonucleotide conjugation and surface modification.

Solubility
Soluble in DCM, DMF, DMSO and THF.

Notes
Moisture sensitive. Incompatible with oxidizing agents and heat.
Specifications

Chemical Identifiers

Azadibenzocyclooctyne-PEG4-NHS ester
649.69
MFCD22380747

Specifications

Azadibenzocyclooctyne-PEG4-NHS ester
649.69
Orange to brown
Moisture Sensitive
Oil
MFCD22380747
25mg
Videos
SDS