missing translation for 'onlineSavingsMsg'
Learn More

Azadibenzocyclooctyne-PEG4-alcohol, Thermo Scientific Chemicals

Catalog Number p-7058247
Click to view available options
Quantity:
25 mg
100 mg
This item is not returnable. View return policy
C29H36N2O7
MFCD22380739
ADIBO-PEG4-alcohol
This item is not returnable. View return policy
C29H36N2O7
MFCD22380739
ADIBO-PEG4-alcohol

ADIBO-containing reagent with PEG4 spacer and alcohol terminus

ADIBO-containing reagent with PEG4 spacer and alcohol terminus

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow
Quantity 25 mg
Formula Weight 524.61
Physical Form Viscous Liquid or Solid
Chemical Name or Material Azadibenzocyclooctyne-PEG4-alcohol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'