Learn More
AZ 3146, Tocris Bioscience™
Potent and selective Mps1 kinase inhibitor
POA
Description
Description
Potent and selective monopolar spindle 1 (Mps1) kinase inhibitor (IC50 = 35 nM). Displays selectivity over 46 other kinases including Cdk1 and aurora kinase B. Interferes with chromosome alignment and overrides spindle assembly checkpoint. Inhibits the recruitment of Mad1, Mad2 and centromere protein E (CENP-E) to kinetochores.