Learn More
Tocris Bioscience™ AZ 10417808
Selective non-peptide caspase-3 inhibitor
Brand:  Tocris Bioscience™ 2172/10
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13440714
  • £183.48 / 10mg
Estimated Shipment
27-06-2019
Description
Description
Selective non-peptide inhibitor of caspase-3 (Ki = 247 nM); displays > 40-fold selectivity over caspases 1, 2, 6, 7 and 8 (Ki > 10 μM). Completely blocks staurosporine-induced intracellular DEVDase activity in SH-SY5Y cells (IC50 = 14.9 μM).
Specifications
Specifications
AZ 10417808
>98
10mg
AZ 10417808
434.23