Learn More
Axygen™ Zymark™ Rapid Plate/Sciclone and Tecan™ TEMO Acquarius Genmate: 50μL
Brand:  Axygen ZT-50-R-S
Code : 68
Additional Details : Weight : 10.89000kg
 View more versions of this product
Product Code. 12716689
  • £263.08 / Pack of 4800
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
  • High throughput screening
  • Volume: 50µL
  • Rack Color: Light Green
  • Color: Clear
  • Designed for use with Zymark® RapidPlate™, Zymark® SciClone, Tecan® Te-MO™, Tecan® Genmate™ and Tecan® Aquarius™ automated liquid handling systems
  • Certified RNase, DNase and Pyrogen-Free
  • Format: 96 well
Specifications
Specifications
Rack
Tecan™ Freedom EVO®þTecan™ TeMoþTecan™ Aquarius MCA96 headþZymark™/Caliper™ RapidPlate™þSciClone™þSciClone™ HV96
Automation
Non-Filtered
Clear
Yes
PP (Polypropylene)
Maxymum Recovery™ to further improve accuracy and precision while reducing sample loss
96 Tips/Rack
Rack
50μL
Sterile
Light Green
Round
Non-ANSI
Tecan™ Freedom EVO®, Tecan™ TeMo, Tecan™ Aquarius MCA96 head, Zymark™/Caliper™ RapidPlate™, SciClone™, SciClone™ HV96, CAS-4200-1ch. 20-150μL/sec. head
Videos
Documentation