Axygen™ Zymark™ Rapid Plate/Sciclone and Tecan™ TEMO Acquarius Genmate: 200μL
Overview
Brand: Axygen
Manufacturer Part Number: ZT-205-WB-R-S
UNSPSC: 41121608
Code: 68
Additional Details:
Additional Details: Weight: 10.90000kg
Product Code. 12766679
Quantity Price
1 £ 246.72 / Pack of 4800
Estimated Shipment
07-04-2017
Add to basket
Description and Specification
Specification
Sterility Sterile
For Use With (Equipment) Tecan™ Freedom Evo™, TeMo and Aquarius with MCA96 head, Zymark™/Caliper RapidPlate™/SciClone™ with HV96 head/CAS-4200-1ch.-20-150μL/sec. head
Quantity 96 Tips/Rack
Material Polypropylene
Product Type Robotic Tip
Autoclavable Yes
Volume (Metric) 200μL
Packaging Rack
Color Clear
Tip Style Racked, Presterilized
Certifications/Compliance Non-ANSI
Shape Wide Bore
Rack Color Blue
Documentation