missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Zymark™ Rapid Plate/Sciclone and Tecan™ TEMO Acquarius Genmate: 200 μL
Clear, autoclavable tips; color-coded racks.
Brand: Axygen™ ZTF-200-L-R-S
Code : 68
Additional Details : Weight : 10.89000kg
Description
- Clear Tips
- Format: 96 well
- Propylene construction
- Certified RNase/DNase free
- Nonpyrogenic
- Aerosol resistant filters
- Sterile
- Compatibility: Tecan™ Freedom Evo™, TeMo and Aquarius with MCA96 head, Zymark™/Caliper RapidPlate™/SciClone™ with HV96 head/CAS-4200-1ch.-20-150μL/sec. head
Specifications
Maxymum Recovery | |
Filtered | |
Clear | |
Autoclavable | |
Maxymum Recovery™ to further improve accuracy and precision while reducing sample loss | |
Automation, Round | |
50 Racks/Cs. | |
Caliper Zymark |
200 μL | |
Sterile | |
Blue | |
Non-ANSI | |
Rack | |
Non-SBS Rack | |
Tecan Freedom EVO, Tecan TeMo, Tecan Aquarius MCA96 head, Zymark/Caliper RapidPlate, SciClone, SciClone HV96, CAS-4200-1ch. 20-150μL/sec. head | |
Polyethylene |