missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Zymark™ Rapid Plate/Sciclone and Tecan™ TEMO Acquarius Genmate: 100 μL
Clear, autoclavable tips; color-coded racks.
Marke: Axygen™ ZT-100-L-R
Code : A0
Weitere Details : Weight : 10.43000kg
Beschreibung
- Tip format: 96 tip and 384 tip
- Tip volume range: 25 μL to 200 μL
- Tip material: Clear polypropylene
- RNase-/DNase-/human gDNA-free
- Nonpyrogenic
- Available options:
- With or without aerosol-resistant filters
- Nonsterile or sterile
- Standard or Axygen™ Maxymum Recovery™ surface - Compatible heads: HV96, CAS-4200-1 (20 - 150 μL/sec.), LV96, LV384
- Rack format: Non-ANSI (96) and ANSI/SLAS (384)
- Rack material: Polypropylene
Bestellinformation
96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
Spezifikation
Maxymum Recovery | |
Non-Filtered | |
Clear | |
Autoclavable | |
Rack | |
Non-SBS Rack | |
Tecan Freedom EVO, Tecan TeMo, Tecan Aquarius MCA96 head, Zymark/Caliper RapidPlate, SciClone, SciClone HV96, CAS-4200-1ch. 20-150μL/sec. head | |
Polypropylene |
100 μL | |
Non-sterile | |
Gray | |
Non-ANSI | |
Automation, Round | |
50 Racks/Cs. | |
Caliper Zymark |