Learn More
Axygen™ Automation Tips for Tecan™, Zymark™, Biotek™ and Dynamic Devices
Ensure consistent performance and quality. Axygen™ Automation Tips for Tecan™, Zymark™, Biotek™ and Dynamic Devices are constructed of clear polypropylene in 96-well racks. Available in nonsterile/sterile, non-filtered/filtered, wide-bore and bottom-labeled options with volumes ranging from 50 to 200μL.
Brand:  Axygen™ EV-50-R-S
Code : 68
Additional Details : Weight : 9.50000kg
 View more versions of this product
Product Code. 12792836
  • £437.97 / Pack of 4800
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
  • Comprehensive line of pipette tips for automated liquid handling systems
  • Manufactured to stringent specifications under strict process controls
  • Assembled via automation to ensure consistent performance and quality
  • Tips are certified RNase-/DNase-free and nonpyrogenic
  • Available in a range of volumes: 50, 100, 180 and 200μL
  • Compatible with the following:
    • Tecan™ Freedom Evo™, TeMo and Aquarius with MCA96 head
    • Zymark™/Caliper RapidPlate™, SciClone™ and Zephyr™ with HV96 head
    • Biotek™ Precision™ and Precision XS 8 or 12 head
    • Dynamic Devices Oasis LM 8/96-Channel-Standard Vol and 96-Channel-Low Vol head
Specifications
Specifications
Tecan™ Freedom EVO®þTecan™ TeMoþTecan™ AquariusþZymark™/Caliper™ RapidPlate™þSciClone™þZephyr™þBioTek™ Precision™þBioTek™ Precision™ XSþDynamic Devices Oasis LM
Automation
Non-Filtered
Clear
Round
ANSI
Non-pyrogenic, used for stringent specifications under strict process controls
Rack
50μL
Sterile
Light Green
PP (Polypropylene)
DNase, Rnase Free
Tecan™ Freedom EVO®, Tecan™ TeMo, Tecan™ Aquarius, Zymark™/Caliper™ RapidPlate™, SciClone™, Zephyr™, BioTek™ Precision™, BioTek™ Precision™ XS, Dynamic Devices Oasis LM
Videos